Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant His6-SARS Virus Papain-like Protease Protein, CF

Catalog No. E610050
Change view
Click to view available options
Quantity: 50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
E610050 Quantity: 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E610050 Supplier R&D Systems™ Supplier No. E610050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Reaction conditions will need to be optimized for each specific application. We recommend an initial PLPro concentration of 20-100 nM when using Ubiquitin-AMC (U-550) or Ubiquitin-Rhodamine 110 (U-555) as a subtrate.

Specifications

Accession Number P0C6U8.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl and TCEP.
Molecular Weight (g/mol) M.W. (theoretical): 37 kDa
Quantity 50 μg
Source E. coli-derived viral Papain-like Protease protein Glu1541 - Lys1855 with a N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Regulatory Status RUO
Common Name Papain-like Protease, PLpro
Product Type Recombinant protein
Conjugate Unconjugated
Recombinant Recombinant
Target Gene SARS Virus Papain-like Protease
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein Papain-like Protease
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.