Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boston Biochem™ Recombinant Human His6-USP16 Protein, CF

Catalog No. E612025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E612025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E612025 Supplier Boston Biochem™ Supplier No. E612025
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Recombinant Human USP16 is a Ubiquitin-specific deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. USP16 does not efficiently utilize Ubiquitin-AMC (U-550), Ubiquitin-Rhodamine 110 (U-555), nor unanchored poly-Ubiquitin chains as substrates in vitro. USP16 cleaves Ubiquitin from recombinant, designer nucleosomes.

Specifications

Accession Number Q9Y5T5
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 7.5, 100 mM NaCl, 10% (v/v) Glycerol, 1 mM TCEP
Gene ID (Entrez) 80824
Molecular Weight (g/mol) M.W. (theoretical): 94 kDa
Quantity 25 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP16 protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Regulatory Status RUO
Common Name MAP kinase phosphatase 7
Product Type Recombinant protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein DUSP16
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.