Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boston Biochem™ Recombinant Human Parkin C431S Protein, CF

Catalog No. E3164100
Change view
Click to view available options
Quantity: 100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
E3164100 Quantity: 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E3164100 Supplier Boston Biochem™ Supplier No. E3164100
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Reaction conditions will need to be optimized for each specific application. As supplied, Parkin C431S has no E3 ligase activity as determined by the lack of autoubiquitination in an in vitro assay.

Specifications

Accession Number O60260
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 25 mM Tris pH 8.5, 200 mM NaCl, 0.03% Brij 35, 10% (v/v) Glycerol, 5 mM TCEP
Gene ID (Entrez) 5071
Molecular Weight (g/mol) M.W. (theoretical): 52 kDa
Quantity 100 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Parkin protein Contains a substitution at Cys431Ser
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Regulatory Status RUO
Gene Alias Parkinson Disease [autosomal recessive, juvenile] 2, Parkin [PARK2], transcript variant 1
Common Name AR-JP, E3 ubiquitin ligase, E3 ubiquitin-protein ligase parkin
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Parkin
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.