Learn More
Boston Biochem™ Human Tetra-Ub (K63-linked) Recombinant Rhodamine 110, CF
Description
Specifications
Specifications
| Accession Number | P0CG47 |
| Formulation | Xmg/mL (XμM) in 50mM HEPES pH 7.0, 50mM NaCl |
| Gene ID (Entrez) | 7314 |
| Molecular Weight (g/mol) | 35 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived human Tetra-Ubiquitin protein |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Regulatory Status | RUO |
| Common Name | TetraUbiquitin |
| Product Type | Recombinant protein |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.