Learn More
R&D Systems™ MgATP Solution
SDP
Catalog No. B20 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
B20 100 μL
1 options
This item is not returnable. View return policy
Catalog No. B20 Supplier R&D Systems™ Supplier No. B20
Add to Cart
Add to Cart
This item is not returnable. View return policy
General Ubiquitin Proteasome Pathway (UPP) Products
Pre-coupled Mg-ATP is an ideal energy source for semi-purified conjugation/degradation reactions. Supplied as a pH adjusted solution with no added buffering reagents.
Stock: 0.1M Mg2+-ATP, pH 7.0
Specifications
Product Line R&D Systems
Quantity 100 μL
Storage Requirements Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.