Learn More
Boston Biochem™ Recombinant Human Di-Ub/Ub2 WT Chains (K27-linked), CF
Description
Specifications
Specifications
| Accession Number | P0CG47 |
| Formulation | 1mg/mL (58μM) in sterile, deionized water |
| Gene ID (Entrez) | 7314 |
| Molecular Weight (g/mol) | 17 kDa |
| Quantity | 25 μg |
| Source | E. coli-derived human Di-Ubiquitin protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Regulatory Status | RUO |
| Common Name | DiUbiquitin |
| Product Type | Recombinant protein |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.