Learn More
Boston Biochem™ Recombinant Human OTUD3 Catalytic Domain Protein, CF
Description
Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human OTUD3 Catalytic Domain concentration of 0.05-0.5 μM.
Specifications
Specifications
| Accession Number | Q5T2D3 |
| For Use With (Application) | Bioactivity |
| Formulation | 1.3 mg/mL (50 μM) in 50 mM HEPES pH 7.5, 100 mM NaCl, 10 % Glycerol (v/v), 1 mM TCEP |
| Gene ID (Entrez) | 23252 |
| Molecular Weight (g/mol) | M.W. (theoretical): 26 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived human OTUD3 protein Glu52 - Gly275 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Gene Symbol | OTUD3 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.