Specifications
| Product Line | TFGlobal,Staged,GID_900000006403154,3747105,ProductLine |
| Quantity | 20 μg |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |