Learn More
Specifications
Specifications
| Form | Lyophilized |
| Product Line | TFGlobal,Staged,GID_900000006403153,3747105,ProductLine |
| Quantity | 20 μg |
| Format | Lyophilized from a solution in deionized water |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
