Learn More
DWK Life Sciences Wheaton™ Boston Round Bottle, Safety Coated, 1000mL
SDP
Catalog No. 02927151 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Liner:
Polyethylene
Polyvinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Liner
02-927-151 Polyvinyl
02-927-154 Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-927-151 Supplier DWK Life Sciences Supplier No. W217853
Add to Cart
Add to Cart
Wheaton clear safety coated Boston Round bottles are manufactured from clear soda lime glass that conforms to USP Type III requirements.
  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Coated glass bottles provide an added measure of safety
  • Case includes hazardous communication label and tamper evident seal if labeling and/or sealing is required
TRUSTED_SUSTAINABILITY
Specifications
Coating Safety Coated
Color Clear
Material Borosilicate Glass (Type 3)
Quantity 12/Cs.
Closure Included Yes
Mouth Wide
Product Type Boston Round Bottle
Closure Liner Polyvinyl
Closure Material Polypropylene