Learn More
DWK Life Sciences Wheaton™ Boston Round Bottle, Safety Coated, Amber, 1000mL
SDP
Catalog No. 02927147 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Liner:
PTFE/Polyethylene
Polyvinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Liner
02-927-147 PTFE/Polyethylene
03-084-186 Polyvinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-927-147 Supplier DWK Life Sciences Supplier No. W217959
Only null left
Add to Cart
Add to Cart
Wheaton safety coated Boston Round bottles are made from Wheaton 900 amber glass that conforms to USP Type III requirements. Amber glass is ideal for protecting light-sensitive samples during storage.
  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Coated glass bottles provide an added measure of safety
  • Case includes hazardous communication label and tamper evident seal if labeling and/or sealing is required
TRUSTED_SUSTAINABILITY
Specifications
Coating Safety Coated
Color Amber
Material Borosilicate Glass (Type 3)
Quantity 12/Cs.
Closure Color White
Closure Included Yes
Mouth Wide
Product Type Boston Round Bottle
Closure Liner PTFE/Polyethylene
Closure Material Polypropylene