Learn More
Promega Bottle for 50% Ethanol for ReliaPrep™ Large Volume HT gDNA Isolation System
Catalog No. PRA2691 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
Catalog No. Quantity
PRA2691 1/Ea.
1 options
Catalog No. PRA2691 Supplier Promega Supplier No. A2691
Reagent bottle for ethanol solution used in automated purification of DNA from 1-10 mL blood on liquid-handling systems.
  • Accessory for use with the ReliaPrep™ Large Volume HT gDNA Isolation System
  • Reagent bottle for ethanol solution used in automated purification of DNA from 1-10mL blood on liquid-handling systems.
Specifications
Quantity 1/Ea.