Learn More
BRANDTECH™ BRAND™ Bottle Adapters for Dispensette™ and seripettor™ Bottletop Dispensers and Titrette™ Bottletop Burettes
Shop All BRANDTECH Scientific ProductsDescription
- Economical polypropylene adapters work for most applications
- ETFE/PTFE adapters for aggressive acids and solvents
- Leading number refers to male thread of adapter
- Second number refers to bottle neck the adapter fits
- 28 mm, 24 mm and 22 mm bottle threads require stacking adapter with 45/33 mm adapter.
Specifications
Specifications
| Material | PTFE, ETFE |
| Neck Type | Bottle Thread: 45 mm |
| Thread Size | External Thread: 33 mm; Bottle Thread: 45 mm |
| Diameter (Metric) Neck | 45 mm |
| Diameter (Metric) Outer Neck | 33 mm |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.