Learn More
Hach Company Carboy, with Spigot, 10-L Capacity
Supplier:  Hach Company 1486858
Encompass_Preferred
Polypropylene with a leak-proof polypropylene screw closure. Molded in spigot provides strength and leak-proof operation. 10 L and 20 L sizes have wide shoulder handles.
Catalog No. 50-199-5142
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.