Learn More
Description
- This large, autoclavable polypropylene bottle can be used for storing and shipping chemicals and reagents.
- The 86 mm diameter opening makes it easy to fill and dispense contents
- Features built-in grips on two sides for secure handling
- Threads should be totally disengaged before autoclaving
Specifications
Specifications
| Capacity (Metric) | 4000 mL |
| Color | Clear |
| Material | Polypropylene |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Threaded |
| Height (Metric) | 290 mm |
| For Use With (Equipment) | Chemicals and Reagents |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

