Learn More
Foxx Life Sciences Autofil PP - Disposable Vacuum Bottle Top Filters for Solvent Filtration, with 0.2um Sterilizing PES Membrane, 500mL, GL45 Thread, Polypropylene Housing, Non-Sterile, 12/CS
Supplier:  Foxx Life Sciences 3251271FLS
Encompass_Preferred
325-1271-FLS: Funnel Only, PP, 500mL, 0.2m PES. Foxx Life Sciences' Autofil PP bottle-top filters for solvent filtration are a revolutionary new way to purify solvents for analytical applications to obtain sterilized or purified extracts. Foxx has developed the world's first single piece polypropylenefunnel, designedto withstand harsh solventsthat would normally degrade traditional bottle-top filter assemblies. All active components are FDA-grade which makes Autofil PP perfect for the extraction of compounds used in the food and drug industry. Foxx Autofil PP is designed to conveniently fit on top of most laboratory bottles with a GL45 thread. Autofil PP is available in 3 different types of membrane: PES, Nylon, and PTFE. It is available in 2 different pore sizes: 0.2m, or 0.45m and it is available in 500mL and 1000mL.
Catalog No. 50-117-8694
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.