Learn More
Thomas Scientific Labmax Dispenser, 5.0 to 25mL
SDP
Supplier:  Thomas Scientific 1185L58
Encompass
Designed Through Research The Labmax™ bottle top dispenser has been developed through extensive testing and input from end users. The result is an easy to use system with a variety of important features. Unique closed circuit air purging system Reagent "guard valve" for safety shut down Exclusive integrated swivel neck Patented direct displacement PTFE piston Quick and accurate volume adjustment Auto-calibrating Fully autoclavable ISO 9001 certified Spill Prevention Feature The unique closed circuit air purging system allows air and bubbles to be eliminated from the dispenser without loss of valuable reagent. The reagent "guard valve" prevents accidental discharge of reagent when the dispenser is not in use. A simple turn of the operations mode selector activates this safety feature. Eliminating the need for discharge tube caps. The exclusive Integrated Swivel Neck allows the discharge tube to be turned 360° for convenient dispensing without affecting the
Catalog No. 50-145-9278
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.