Learn More
Corning™ Bottle Top Dispenser (BTD) PTFE Plunger
Catalog No. 10320162 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
10-320-162 2.5 mL
10-320-163 5 mL
10-320-164 10 mL
10-320-165 25 mL
10-320-166 50 mL
10-320-167 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 10-320-162 Supplier Corning™ Supplier No. 6861
Add to Cart
Add to Cart
This item is not returnable. View return policy
PTFE plunger for BTD and dispensing reagents
  • Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
  • Comes with four adapters with different diameters for universal use with multiple bottle sizes
  • Telescopic suction tube can be adjusted smoothly to different bottle heights
  • Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
  • Every dispenser is individually tested and delivered with a certificate of performance
  • Uses:
    • For dispensing organic and inorganic liquids
  • TRUSTED_SUSTAINABILITY
    Specifications
    Volume (Metric) 2.5 mL
    For Use With (Application) Dispensing Organic and Inorganic Liquids
    Includes Four adapters