Learn More
Corning™ Bottle Top Dispensers (BTD) Outer Housing
Catalog No. 10320151 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2.5 mL
5 mL
25 mL
50 mL
100 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
10-320-151 100 mL
10-320-147 2.5 mL
10-320-148 5 mL
10-320-149 25 mL
10-320-150 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10-320-151 Supplier Corning™ Supplier No. 6850
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Outer housing for BTD and dispensing reagents
  • Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
  • Comes with four adapters with different diameters for universal use with multiple bottle sizes
  • Telescopic suction tube can be adjusted smoothly to different bottle heights
  • Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
  • Every dispenser is individually tested and delivered with a certificate of performance
  • Uses:
    • For dispensing organic and inorganic liquids
  • TRUSTED_SUSTAINABILITY
    Specifications
    Volume (Metric) 100 mL
    For Use With (Application) Dispensing Organic and Inorganic Liquids
    Includes Four adapters