Learn More
Hach Company Bottle, Sampling, Polycarbonate, 125 mL, 48/pk
Supplier:  Hach Company 2324373
Encompass_Preferred
Transparent polycarbonate bottle with cap features wide mouth (3.8cm/1.48" diameter) opening for easy filling and square design for maximum space efficiency. Autoclavable with molded-in graduations (do not engage threads for autoclave cycle.)
Catalog No. 50-199-5807
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.