Learn More
Hach Company Bottle, Wash, Polyethylene, Narrow Mouth, 125 mL, 12/pk
Supplier:  Hach Company 2930101
Encompass_Preferred
Polyethylene bottle with polypropylene closure and stem molded in one piece for leak-proof service. Tip can be cutback to increase flow. Available in Narrow or Wide Mouth.
Catalog No. 50-199-6834
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.