Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Bottles, Storage, Wide Mouth
SDPSCI_ED

Catalog No. S15744 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S15744 500 mL
S15757 250 mL
S15940 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S15744 Supplier United Scientific™ Supplier No. 81901
Add to Cart
Add to Cart

Safely store liquids and powders.

  • These bottles provide an excellent option for storage of reagents and other materials in the laboratory.
  • The square shape is designed to optimize shelf space.
  • Available in three different sizes, these bottles feature polypropylene caps, are autoclavable, and provide good chemical resistance.

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Color Clear
Material Polypropylene
Quantity 12/Pk.
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Threaded
Shape Square
Height (Metric) 115 mm
Product Type Square Wide Mouth Bottle
For Use With (Equipment) Lab Scoops
Closure Material Polypropylene
Closure Size 49 mm ID
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.