Learn More
Description
- These bottles provide an excellent option for storage of reagents and other materials in the laboratory.
- The square shape is designed to optimize shelf space.
- Available in three different sizes, these bottles feature polypropylene caps, are autoclavable, and provide good chemical resistance.
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Color | Clear |
| Material | Polypropylene |
| Quantity | 6/Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Threaded |
| Height (Metric) | 135 mm |
| For Use With (Equipment) | Lab Scoops |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

