Learn More
MilliporeSigma™ Supelco™ Bottom-drip adapter for large Columns Luer tip, 1 mm bore, PTFE
Catalog No. 111009597 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
11-100-9597 PTFE
1 options
Catalog No. 11-100-9597 Supplier MilliporeSigma™ Supelco™ Supplier No. Z1801731EA
Add to Cart
Add to Cart
Shallow taper of upper end minimizes mixing below the PE packing support disc (included). All adapters have an Ace Thred No. 50 and include one FETFE™ O-ring.
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE
FETFE is a registered trademark of Ace Glass, Inc.