Learn More
MilliporeSigma™ Supelco™ Bottom-drip adapter for large Columns Luer tip and flow-regulator valve, 0 to 1 mm adjustable bore, Nylon
Catalog No. 111009765 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Nylon
Catalog No. Material
11-100-9765 Nylon
1 options
Catalog No. 11-100-9765 Supplier MilliporeSigma™ Supelco™ Supplier No. Z1817061EA
Shallow taper of upper end minimizes mixing below the PE packing support disc (included). All adapters have an Ace Thred No. 50 and include one FETFE™ O-ring.
TRUSTED_SUSTAINABILITY
Specifications
Material Nylon
FETFE is a registered trademark of Ace Glass, Inc.