Learn More
MilliporeSigma™ Supelco™ Bottom-drip adapter for large Columns Luer tip and flow-regulator valve, 0 to 1 mm adjustable bore, PTFE
Catalog No. 111009240 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
11-100-9240 PTFE
1 options
Catalog No. 11-100-9240 Supplier MilliporeSigma™ Supelco™ Supplier No. Z1801811EA
Add to Cart
Add to Cart
Shallow taper of upper end minimizes mixing below the PE packing support disc (included). All adapters have an Ace Thred No. 50 and include one FETFE™ O-ring.
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE
FETFE is a registered trademark of Ace Glass, Inc.