Learn More
Description
- Provide a particulate barrier and helps prevent hair from contaminating a product or work environment
- Low-linting, lightweight cover restrains hair and offers great ventilation
- Designed to protect against dirt, grime, and particulates in non-hazardous environments
- Flat style
- Latex-free elastic
- Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
Specifications
| Product Type | Bouffant |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Model | S110NW |
| Diameter (English) | 21 in. |
| For Use With (Application) | Food Processing, Construction, Pharmaceuticals, Environmental Cleanup |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.