Description
- Provide a particulate barrier and helps prevent hair from contaminating a product or work environment
- Low-linting, lightweight cover restrains hair and offers great ventilation
- Designed to protect against dirt, grime, and particulates in non-hazardous environments
- Flat style
- Latex-free elastic
- Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
Specifications
| Product Type | Bouffant |
| Material | Polypropylene |
| Disposable | Single-use |
| Model | S110NW |
| Diameter (English) | 21 in. |
| For Use With (Application) | Food Processing, Construction, Pharmaceuticals, Environmental Cleanup |