Learn More
Sigma Aldrich Fine Chemicals Biosciences Bouins solution
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HT1011284L
Encompass
Bouin SOLUTIONon is a noncoagulant yellow stain fixative. It imposes shrinkage on tissues when used as preservative due to its picric acid component. It is majorly used in ureteroscopic biopsies. It preserves morphological and nuclear information in tissues upon fixation.
Catalog No. 50-290-4812
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.