Learn More
PIP™ Bouton™ Bullseye™ Rimless Safety Glasses
Catalog No. 19153341 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Anti-reflective, Scratch-resistant
Scratch-resistant
Lens Tint:
Clear
Gradient
Gray
Light Blue
Frame Color:
Clear
Gray
Light Blue
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint Frame Color
19-153-341 Scratch-resistant Gray Gray
19-153-344 Anti-reflective, Scratch-resistant Clear Clear
19-153-342 Scratch-resistant Light Blue Light Blue
19-153-347 Scratch-resistant Gradient Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-153-341 Supplier PIP™ Supplier No. 250310021
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Trendy design that hits the mark with a lens treatment to fit your application.
  • Lightweight, rimless design offers optimal protection
  • Flexible bayonet style temples provide a secure fit
  • Molded nose bridge design to fit the vast majority of users
  • Gray Lens - visible light transmission of 12% and a darkening effect with little color distortion, best used outdoors in bright sun
  • Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
  • Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Specifications
Lens Coating Scratch-resistant
Lens Tint Gray
Product Type Safety glasses
Frame Color Gray
Lens Material Polycarbonate