Description
- Lightweight, rimless design offers optimal protection
- Flexible bayonet style temples provide a secure fit
- Molded nose bridge design to fit the vast majority of users
- Gradient tint lenses have a slight mirror coating on the top portion of the lens which reduces glare from high-powered overhead fluorescent and mercury lighting helping to minimize squinting and eye strain, resulting in increased productivity
- The light yellow tint blocks the harsh blue light emitted from electronic screens and the slight mirror coating on the outside reduces glare from overhead lighting
- Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
- Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Lens Tint | Gradient |
| Product Type | Safety glasses |
| Frame Color | Clear |
| Lens Material | Polycarbonate |