Learn More
PIP™ Bouton™ Captain™ Rimless Safety Glasses
Catalog No. 19153359 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Clear
Gray
Lens Coating:
Anti-fog, Scratch-resistant
Anti-reflective, Scratch-resistant
Lens Tint:
Blue, Indoor/Outdoor
Clear
Gray
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-153-359 Clear Anti-fog, Scratch-resistant Blue, Indoor/Outdoor
19-153-358 Clear Anti-reflective, Scratch-resistant Clear
19-153-356 Clear Anti-fog, Scratch-resistant Clear
19-153-357 Gray Anti-fog, Scratch-resistant Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-153-359 Supplier PIP™ Supplier No. 250460226
Add to Cart
Add to Cart
This item is not returnable. View return policy
Popular one-piece lens design that offers a perfect fit.
  • Lightweight, rimless design provides all day user comfort
  • Bayonet temples with end tips that curve out for ease of placement on face
  • Non-slip rubber nose pads offer a secure fit
  • Clear Lens for clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
  • Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
  • Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Blue, Indoor/Outdoor
Product Type Safety Glasses
Frame Color Clear
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Frameless
Frame Type Frameless
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.