Learn More
PIP™ Bouton™ Captain™ Rimless Safety Glasses
Catalog No. p-4817280 Shop All Protective Industrial Products Inc. Products
Click to view available options
Frame Color:
Clear
Gray
Lens Coating:
Anti-fog, Scratch-resistant
Anti-reflective, Scratch-resistant
Lens Tint:
Blue, Indoor/Outdoor
Clear
Gray
This item is not returnable. View return policy
This item is not returnable. View return policy
Popular one-piece lens design that offers a perfect fit.
  • Lightweight, rimless design provides all day user comfort
  • Bayonet temples with end tips that curve out for ease of placement on face
  • Non-slip rubber nose pads offer a secure fit
  • Clear Lens for clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
  • Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
  • Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Specifications
Lens Coating Anti-reflective, Scratch-resistant
Lens Tint Clear
Product Type Safety Glasses
Frame Color Clear
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Frameless
Frame Type Frameless
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.