Learn More
PIP™ Bouton™ Commander™ Semi-Rimless Safety Glasses
Catalog No. 19153377 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Gloss Black
Gloss Bronze
Lens Coating:
Anti-fog, Scratch-resistant
Anti-reflective, Scratch-resistant
Scratch-resistant
Lens Tint:
Blue, Indoor/Outdoor
Brown
Clear
Gray
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-153-377 Gloss Bronze Anti-fog, Scratch-resistant Blue, Indoor/Outdoor
19-153-369 Gloss Black Anti-fog, Scratch-resistant Clear
19-153-370 Gloss Black Anti-fog, Scratch-resistant Gray
19-153-372 Gloss Black Anti-fog, Scratch-resistant Blue, Indoor/Outdoor
19-153-373 Gloss Black Scratch-resistant Gray
19-153-376 Gloss Bronze Scratch-resistant Clear
19-153-374 Gloss Bronze Anti-fog, Scratch-resistant Clear
19-153-375 Gloss Bronze Anti-fog, Scratch-resistant Brown
19-153-371 Gloss Black Anti-reflective, Scratch-resistant Clear
19-153-378 Gloss Bronze Scratch-resistant Brown
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-153-377 Supplier PIP™ Supplier No. 250331226
Add to Cart
Add to Cart
This item is not returnable. View return policy
Unique styling that will command your attention with fit, comfort and a variety of technical lens treatments.
  • Lightweight, semi-rimless design offers extensive eye coverage and all day comfort
  • Bayonet style temples with non-slip rubber pads provide a secure fit
  • Adjustable rubber nose pads offer a custom, comfortable fit
  • Clear Lens for clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
  • Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
  • Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Blue, Indoor/Outdoor
Product Type Safety Glasses
Frame Color Gloss Bronze
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
Frame Type Semi-rimless
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.