Learn More
PIP™ Bouton Contempo Goggles
Catalog No. 19148214 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Includes:
Wide elastic headband with adjustable buckle
Wide neoprene headband with adjustable buckle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Includes
19-148-214 Scratch-resistant Wide elastic headband with adjustable buckle
19-148-215 Anti-fog, Scratch-resistant Wide neoprene headband with adjustable buckle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-148-214 Supplier PIP™ Supplier No. 2515300000
Add to Cart
Add to Cart
This item is not returnable. View return policy
PIP™ Bouton Contempo Goggles feature a wraparound style that offers a snug, comfortable fit with a wide viewing area.
  • Wraparound lens design with distortion-free panoramic viewing
  • Ideal for general manufacturing and anywhere eye protection is needed.
  • Offers indirect ventilation for liquid splash protection
  • Wide facial flange for a secure seal
  • Nose area split to fit the vast majority of users
  • Clear blue transparent frame to prevent yellowing
  • Fits comfortably over most prescription glasses
  • Headband strap attaches to frame with a swivel clip to ensure a comfortable fit
  • 6 Base curve lens
  • Meets ANSI Z87.1
Specifications
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Indirect Vent Safety Goggles
Frame Color Light Blue
Includes Wide elastic headband with adjustable buckle
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material PVC
Frame Type Indirect Vented
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Warning - Prop 65 - Reproductive Harm