Learn More
PIP™ Bouton™ Fuselage™ Full Frame Safety Glasses
Shop All Protective Industrial Products Inc. ProductsDescription
- Flame resistant soft foam padding forms a seal to keep out dust and airborne particles
- Comes with both temples AND elastic headband strap
- Temples and strap snap in or out - easily converts from a safety glass to a dust goggle and back
- Bayonet nylon temples with rubber tips for a secure fit
- Molded nose bridge design to fit the vast majority of users
- Clear Lens for clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
- Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
- Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Amber |
| Product Type | Safety Glasses |
| Frame Color | Black, Gray |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| UV Protection | 99.9% UVA/UVB/UVC |
| Frame Type | Sealed with Foam |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.