Learn More
PIP™ Bouton™ Radar™ Rimless Safety Glasses
Catalog No. 19153355 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Anti-reflective, Scratch-resistant
Lens Tint:
Blue, Indoor/Outdoor
Clear
Gray
Frame Color:
Gray
Red
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint Frame Color
19-153-355 Anti-fog, Scratch-resistant Blue, Indoor/Outdoor Red
19-153-349 Anti-fog, Scratch-resistant Gray Gray
19-153-352 Anti-fog, Scratch-resistant Clear Red
19-153-353 Anti-fog, Scratch-resistant Gray Red
19-153-348 Anti-fog, Scratch-resistant Clear Gray
19-153-354 Anti-reflective, Scratch-resistant Clear Red
19-153-351 Anti-fog, Scratch-resistant Blue, Indoor/Outdoor Gray
19-153-350 Anti-reflective, Scratch-resistant Clear Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-153-355 Supplier PIP™ Supplier No. 250451226
Add to Cart
Add to Cart
This item is not returnable. View return policy
One-piece lens design with stylish temples that are sure to get you noticed.
  • Lightweight, rimless design offers optimal protection
  • Flexible bayonet style temples provide a secure fit
  • Non-slip rubber nose pads provide a secure fit
  • Clear Lens for clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
  • Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
  • Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Blue, Indoor/Outdoor
Product Type Safety Glasses
Frame Color Red
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection 99.9% UVA/UVB/UVC
Frame Type Rimless
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.