Learn More
PIP™ Bouton™ Softsides Cover Chemical Splash Goggles
Catalog No. 19052558 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Uncoated
Catalog No. Lens Coating
19-052558 Uncoated
1 options
This item is not returnable. View return policy
Catalog No. 19-052558 Supplier PIP™ Supplier No. 2485190300B
  • $17.00
    $11.65 / Pair
    Save $5.35
    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $2,098.00
    $1,458.65 / Case of 144 PR
    Save $639.35
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
For chemical splash protection
  • Clear polycarbonate lens
  • Soft vinyl body fits over most prescription eyewear and molds comfortably to facial contours
  • Added ribs reinforce body for lens retention and superior impact resistance
  • With elastic headband
  • Hooded vents for indirect ventilation
Order Info
Available in standard or Fogless™ versions.
Certifications
CSA standards.
Compliance
Chemical splash goggle meets or exceeds ANSI™ Z87.1-1989
Specifications
Lens Coating Uncoated
Certifications/Compliance ANSI Z87.1-1989, CSA standards
Lens Tint Clear
Product Type Over-the-Glasses Cover Goggles
For Use With (Equipment) Fits over most prescription eyewear
For Use With (Application) Chemical splash protection
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Vinyl
Frame Type Indirect Vented