Learn More
PIP™ Bouton™ Softsides Cover Impact Goggles
Catalog No. 19052556 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Product Type:
Over-the-Glasses Cover Goggles
Lens Coating:
Anti-fog, Scratch-resistant
Uncoated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Lens Coating
19-052556 Over-the-Glasses Cover Goggles Anti-fog, Scratch-resistant
19-052552 Over-the-Glasses Cover Goggles Uncoated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-052556 Supplier PIP™ Supplier No. 2485090400B
  • $11.50
    $8.65 / Each
    Save $2.85
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,438.00
    $1,083.65 / Case of 144 EA
    Save $354.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
For impact protection
  • Clear polycarbonate lens
  • Soft vinyl body fits over most prescription eyewear and molds comfortably to facial contours
  • Added ribs reinforce body for lens retention and superior impact resistance
  • With elastic headband
  • Perforations in frame allow constant airflow for direct ventilation
Order Info
Available in standard or Fogless™ versions.
Certifications
CSA standards.
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1-1989, CSA standards
Lens Tint Clear
Product Type Over-the-Glasses Cover Goggles
For Use With (Equipment) Fits over most prescription eyewear
For Use With (Application) Impact protection
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Vinyl
Frame Type Direct Vent
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.