Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine

Catalog No. MP210129125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210129125 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210129125 Supplier MP Biomedicals™ Supplier No. 0210129125
Add to Cart
Add to Cart

Used as a microbiological nutrient in laboratory media and fermentations.

Preparation Method: Product is prepared by acid digestion of casein.
Applications: Casein acid hydrolysate is used as a microbiological nutrient in laboratory media and fermentations. It has also been used as a nutrient in vaccine and toxin production where a high free amino acid content is required.
Product Description: Casein, Acid Hydrolysate is a hydrochloric acid hydrolysate of casein for use in preparing microbiological culture media. Casein is a rich source of amino nitrogen. Treatment with hydrochloric acid hydrolyzes the protein into primarily free amino acids with some short peptide fragments. Tryptophan and vitamins present in the casein are destroyed by the acid hydrolysis. The salt content of this peptone is very high at approximately 36%.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow/Cream
pH 5.0 to 7.0 (2% aq soln)
For Use With (Application) Media Components
Content And Storage Store at Room Temperature (15-30 °C).
Quantity 25 lb.
Product Type Proteins, Enzymes & Peptides
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.