Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Albumin Cohn Modified, Lyophilized

Catalog No. MP08840042 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08840042 50 g
MP08840043 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP08840042 Supplier MP Biomedicals™ Supplier No. 08840042
Add to Cart
Add to Cart

Used to solubilize lipids, as blocking agent in Western blots or ELISA applications.

  • Cohn modified, heat-shocked fractionated
TRUSTED_SUSTAINABILITY

Specifications

Concentration Protein: >96%
For Use With (Application) Immunoassay, Mammalian Cell Culture
pH Range 6.84
Quantity 50 g
Source Bovine
Storage Requirements Store at 2° to 8°C
Product Type Bovine albumin
Form Powder
Protein Subtype Albumins
Purity or Quality Grade >96%By Protein Electrophoresis (Albumin Fraction V)

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.