Learn More
Haematologic Technologies Inc BOVINE ALPHA THROMBIN 1MG
Supplier:  Haematologic Technologies Inc BCT10201MG
Encompass_Preferred
Coagulation Protein, Bovine alpha-Thrombin, 1 milligram, Native enzyme, Mode of action: Serine protease which cleaves fibrinogen to form fibrin, Research use only, 50% (v/v) glyercol/water, store at -20 Celsius
Catalog No. 50-883-393
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.