Learn More
BIOHIPPO BOVINE ANX-1 ELISA KIT 96T
SDP
Supplier:  BIOHIPPO BHE12100286
Encompass
Bovine annexinⅠ (ANX-Ⅰ) ELISA Kit 96T. This sandwich kit is for the accurate quantitative detection of AnnexinⅠ (also known as ANX-Ⅰ) in serum, plasma, cell culture supernates, cell lysates, tissue homogenates.
Catalog No. NC2584883
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.