Learn More
Biomatik Corporation Bovine Casein Alpha (CSN1) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10912
Encompass_Preferred
Bovine Casein Alpha (CSN1) High-sensitive ELISA Kit for detecting CSN1 in milk. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02662. Gene ID: 282208. Target Synonyms: CASa; CSN1; CSN1S1; Casein Alpha S1; Caseinate; Caseine/caséine/Kasein/caseína; Alpha-S1-casein; Casoxin-D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5288
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.