Learn More
Kingfisher Biotech Inc Bovine CD27 Recombinant Protein
Supplier: Kingfisher Biotech Inc RP1718B005
The Bovine CD27 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine CD27 applications are for cell culture, ELISA standard, and Western Blot Control. Bovine CD27 yeast-derived recombinant protein can be purchased in multiple sizes. Bovine CD27 Specifications: (Molecular Weight: 17.7 kDa) (Amino Acid Sequence: TPGPKSCPEK HYWAQGGWCC QMCEPGTFLV KDCEQHREAA QCNPCTPGVS FMPDHHSRPH CESCRHCNSG LLIRNCTLTA NSECACPEGQ QCRDKDCMEC DGPAQAPGPH PQPSHLPYAE EIAESRTDRH TQTLANSRWL PAPTLSTHWS PQRSLCSANC (160)) (Gene ID: 512514). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.