Learn More
Eurofins Discoverx Bovine Type II Collagen, Immunization Grade, 10 mg. (92-1090)
Supplier: Eurofins Discoverx 921090
Type II collagen makes up 50% of all protein in cartilage, and is particularly prevalent in articular and hyaline cartilage. Forms fibrils that allows cartilage to trap proteoglycan aggregate and provide tissue tensile strength. This product contains bovine-derived collagen suitable for immunization to induce development of arthritis in animal models. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter, PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.