Learn More
Kingfisher Biotech Inc Bovine FGF basic Recombinant Protein
Supplier: Kingfisher Biotech Inc RP1304B005
The Bovine FGF basic yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine FGF basic applications are for cell culture, ELISA standard, and Western Blot Control. Bovine FGF basic yeast-derived recombinant protein can be purchased in multiple sizes. Bovine FGF basic Specifications: (Molecular Weight: 16.3 kDa) (Amino Acid Sequence: SLPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY SSWYVALKRT GQYKLGPKTG PGQKAILFLP MSAKS (145)) (Gene ID: 281161). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.