Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine FGF basic/FGF2/bFGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB33NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB33NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB33NA Supplier R&D Systems Supplier No. AB33NA
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 4 publications

FGF basic/FGF2/bFGF Polyclonal specifically detects FGF basic/FGF2/bFGF in Bovine samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen FGF basic/FGF2
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 1.5-4.5 ug/mL
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Gene Symbols FGF2
Host Species Rabbit
Immunogen Bovine brain-derived FGF basic/FGF2
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2247
Test Specificity Detects bovine FGF basic/FGF2 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with FGF acidic and recombinant human (rh) beta-ECGF is observed and less than 1% cross-reactivity with rhFGF-4, rhFGF-5, rhFGF-6, rhFGF-7, recombinant mouse (rm) FGF-8b, rmFGF-8c, rhFGF-9, rhFGF-10, rmFGF-15, rhFGF-17, and rhFGF-18 is observed. Neutralizes the biological activity of bovine FGF basic/FGF2 and will also neutralize the biological activity of recombinant human FGF basic/FGF2.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Bovine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.