Learn More
American Research Products Inc BoVine Fibrinogen Fb Elisa Ki
Supplier:  American Research Products Inc CSBE13283B
Encompass
BoVine Fibrinogen Fb Elisa Kit from CUSABIO Detection Range 3 125 g/mL-200 g/mL Sensitivity 1 56 g/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26633
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.