Learn More
Bioworld Bovine Gamma Globulin Lyophilized Powder, ≥ 96%, 10 g
Supplier:  Bioworld 220700252
Encompass_Preferred
Purity levels as GG are analyzed by SDS-Page or Agarose electrophoresis. These preparations are especially useful as a co-precipitant in RIA methods employing polyethylene glycol or ammonium sulfate precipitation steps, or as a protein stabilizer, or as blocker in a number of immunoassays. These preparations are also useful as GG control solutions for biochemical analytical tests. Gamma globulin preparations reconstitute to pH 7.2 ± 0.2.Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.
Catalog No. 50-253-941
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.